missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Integrin beta 4/CD104 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB4054N-100UG
This item is not returnable.
View return policy
Description
Integrin beta 4/CD104 Monoclonal specifically detects Integrin beta 4/CD104 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Integrin beta 4/CD104 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_001005608 | |
| ITGB4 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin beta 4/CD104 Asn29-Ser711 Accession # NP_001005608 | |
| RUO | |
| 3691 | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 308601 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD104, CD104 antigen, GP150, integrin beta-4, integrin beta-4 subunit, integrin, beta 4, ITGB4 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse Integrin beta 4/CD104 in direct ELISAs and Western blots. Does not cross-react with recombinant mouse Integrin beta 1, beta 2, beta 6, beta 7, or recombinant human Integrin beta 3. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction