missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-17F Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems IC2057U-100UG
This item is not returnable.
View return policy
Description
IL-17F Monoclonal specifically detects IL-17F in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IL-17F | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q7TNI7 | |
| IL17F | |
| E. coli-derived recombinant mouse IL-17F Arg29-Ala161 Accession # Q7TNI7 | |
| RUO | |
| 112744 | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 316016 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Cytokine ML-1, IL17F, IL-17Finterleukin-17F, interleukin 17F | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse IL-17F in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) IL-17, rmIL-17B, C, D, E, or recombinant human IL-17F is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction