missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Wnt-9a Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC3157T-100UG
This item is not returnable.
View return policy
Description
Wnt-9a Monoclonal specifically detects Wnt-9a in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Wnt-9a | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| O14904.2 | |
| WNT9A | |
| E. coli-derived recombinant human Wnt-9a Pro48-Glu92 & Arg231-Phe304 Accession # O14904.2 | |
| RUO | |
| 7483 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 326711 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| MGC138165, MGC141991, Protein Wnt-14, wingless-type MMTV integration site family, member 14, wingless-type MMTV integration site family, member 9A, Wnt-14, WNT14protein Wnt-9a, Wnt9a | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Wnt-9a in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Wnt-2, -3a, -6, -7a, -7b, recombinant mouse Wnt-1, -2b, -4, -5a, -5b, -8a, -8b, -9b, -10a, -10b, -11, or -16 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction