missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TLX/NR2E1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems PP-H6506-00
This item is not returnable.
View return policy
Description
TLX/NR2E1 Monoclonal specifically detects TLX/NR2E1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Direct ELISA.
Specifications
| TLX/NR2E1 | |
| Monoclonal | |
| Unconjugated | |
| A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide | |
| NR2E1 | |
| Recombinant human TLX (amino acids 81 - 180) and mouse myeloma cells (NS-1) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation, ELISA | |
| H6506 | |
| Western Blot, Immunoprecipitation, Direct ELISA | |
| hTll, NR2E1, nuclear receptor subfamily 2, group E, member 1, Nuclear receptor TLX, Protein tailless homolog, tailes-related receptor, tailless homolog, TLL, TLXnuclear receptor subfamily 2 group E member 1, XTLL | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7101 | |
| Store the unopened product at -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction