missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TIM-4 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB2929V-100UG
This item is not returnable.
View return policy
Description
TIM-4 Monoclonal specifically detects TIM-4 in Human samples. It is validated for Flow Cytometry.
Specifications
| TIM-4 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| FLJ27515, SMUCKLER, T-cell immunoglobulin and mucin domain containing 4, T-cell immunoglobulin and mucin domain-containing protein 4, T-cell membrane protein 4, TIM4, TIM-4, TIMD4, TIMD-4 | |
| Mouse myeloma cell line NSO-derived recombinant human TIM-4 Glu25-Leu315 Accession # Q96H15 | |
| RUO | |
| 91937 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 921832 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q96H15 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human TIM-4 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction