missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TACE/ADAM17 Cytosolic Domain Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB21291V-100UG
This item is not returnable.
View return policy
Description
TACE/ADAM17 Monoclonal specifically detects TACE/ADAM17 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| TACE/ADAM17 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P78536 | |
| ADAM17 | |
| E. coli-derived recombinant human TACE/ADAM17 Asp695-Cys824 Accession # P78536 | |
| 100 μg | |
| Primary | |
| Detects human TACE/ADAM17 Cytosolic Domain in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human ADAM33 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
| Flow Cytometry | |
| 136133 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| ADAM 17, ADAM metallopeptidase domain 17, ADAM metallopeptidase domain 18, ADAM17, ADAM18, CD156b, CD156b antigen, CSVP, disintegrin and metalloproteinase domain-containing protein 17, EC 3.4.24.86, MGC71942, Snake venom-like protease, TACEcSVP, TNF-alpha convertase, TNF-alpha converting enzyme, TNF-alpha-converting enzyme, tumor necrosis factor, alpha, converting enzyme | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6868 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction