missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NRAGE Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB3835V-100UG
This item is not returnable.
View return policy
Description
NRAGE/MAGED1 Monoclonal specifically detects NRAGE/MAGED1 in Human samples. It is validated for Flow Cytometry.
Specifications
| NRAGE/MAGED1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9Y5V3 | |
| MAGED1 | |
| E. coli-derived recombinant human NRAGE Met1-Glu778 Accession # Q9Y5V3 | |
| RUO | |
| 9500 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 353124 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| DLXIN-1, MAGE tumor antigen CCF, MAGED1, melanoma antigen family D, 1, melanoma-associated antigen D1, Neurotrophin receptor-interacting MAGE homolog, NRAGE, NRAGE MAGE-D1 antigen | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human NRAGE in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction