missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MMP-25/MT6-MMP Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB11422N-100UG
This item is not returnable.
View return policy
Description
MMP-25/MT6-MMP Monoclonal specifically detects MMP-25/MT6-MMP in Human samples. It is validated for Flow Cytometry.
Specifications
| MMP-25/MT6-MMP | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9NPA2 | |
| MMP25 | |
| Mouse myeloma cell line NS0-derived recombinant human MMP-25/MT6-MMP Pro314-Cys508 Accession # Q9NPA2 | |
| RUO | |
| 64386 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 141811 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| leukolysin, matrix metallopeptidase 25, matrix metallopeptidase-like 1, matrix metalloproteinase 20, matrix metalloproteinase 25, matrix metalloproteinase-25, matrix metalloproteinase-like 1, membrane-type 6 matrix metalloproteinase, membrane-type matrix metalloproteinase 6, membrane-type-6 matrix metalloproteinase, MMP20, MMP20A, MMP25, MMP-25, MMPL1, MT6MMP, MT6-MMP, MT-MMP 6, MTMMP6, MT-MMP6 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human MMP-25/MT6-MMP in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction