missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MINA Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC7476T-100UG
This item is not returnable.
View return policy
Description
MINA Monoclonal specifically detects MINA in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| MINA | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q8IUF8 | |
| MINA | |
| E. coli-derived recombinant human MINA Met1-Gly192 Accession # Q8IUF8 | |
| RUO | |
| 84864 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 753002 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| DKFZp762O1912, FLJ14393, MDIG, MDIGDKFZp762O1912, Mina53, MINA53mdig, mineral dust induced gene protein, Mineral dust-induced gene protein, MYC induced nuclear antigen, MYC-induced nuclear antigen, myc-induced nuclear antigen, 53 kDa, NO52, Nucleolar protein 52 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human MINA in direct ELISAs. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse MINA is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction