missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Melanocortin-5 R/MC5R Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB8205S-100UG
This item is not returnable.
View return policy
Description
Melanocortin-5 R/MC5R Monoclonal specifically detects Melanocortin-5 R/MC5R in Human samples. It is validated for Flow Cytometry.
Specifications
| Melanocortin-5 R/MC5R | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P33032 | |
| MC5R | |
| NS0 mouse myeloma cell line transfected with human Melanocortin-5 R/MC5R Met1-Asp325 Accession # P33032 | |
| RUO | |
| 4161 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 821012 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| MC2, MC-2, MC5, MC5R, MC5-R, melanocortin 5 receptor, melanocortin receptor 5, melanocortin-5 receptor, Melanocortin5R, Melanocortin-5R | |
| Mouse | |
| 100 μg | |
| Primary | |
| Stains human Melanocortin-5 R/MC5R transfected cells but not irrelevant transfectants in flow cytometry. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction