Learn More
Human LBP Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB870R-100
 View more versions of this product
Product Code. 18714829
  • £388.00 / 100µg
Description
LBP Monoclonal specifically detects LBP in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LBP
Monoclonal
Unconjugated
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
lipopolysaccharide binding protein, lipopolysaccharide-binding protein, LPS-binding protein, MGC22233
Mouse myeloma cell line NS0-derived recombinant human LBP Ala26-Val481 Accession # AAD21962
100 μg
3929
Reconstitute at 0.5mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1
ELISA
127408R
ELISA
AAD21962
Mouse
Protein A or G purified from cell culture supernatant
Primary
Detects human LBP in direct ELISAs.
Human
Supernatant