missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human KIR2DL1/KIR2DS5 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB1844T-100UG
This item is not returnable.
View return policy
Description
KIR2DL1/KIR2DS5 Monoclonal specifically detects KIR2DL1/KIR2DS5 in Human samples. It is validated for Flow Cytometry.
Specifications
| KIR2DL1/KIR2DS5 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P43626 | |
| KIR2DL1 | |
| BaF3 mouse pro-B cell line transfected with human KIR2DL1/KIR2DS5 Accession # P43626 | |
| RUO | |
| 3802 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 143211 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD158a, CD158g, NKAT9 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human KIR2DL1/KIR2DS5 on transfected cells. It does not stain cells transfected with KIR2DL2, 2DL3, 2DL4, 2DL5, 2DS1, 2DS2, 2DS4, 3DL1, 3DL2, or 3DS1. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction