missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GDF-9 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB8266T-100UG
This item is not returnable.
View return policy
Description
GDF-9 Monoclonal specifically detects GDF-9 in Human samples. It is validated for Flow Cytometry.
Specifications
| GDF-9 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| GDF9, GDF-9, growth differentiation factor 9, growth/differentiation factor 9 | |
| Human embryonic kidney cell line derived recombinant human GDF-9 Met1-Arg454 Accession # O60383 | |
| RUO | |
| 2661 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 917319 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| O60383 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human GDF-9 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction