missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DEP-1/CD148 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB19341R-100UG
This item is not returnable.
View return policy
Description
DEP-1/CD148 Monoclonal specifically detects DEP-1/CD148 in Human samples. It is validated for Flow Cytometry.
Specifications
| DEP-1/CD148 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q12913 | |
| PTPRJ | |
| E. coli-derived recombinant human DEP-1/CD148 Arg997-Ala1337 Accession # Q12913 | |
| RUO | |
| 5795 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 261922 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD148, CD148 antigen, Density-enhanced phosphatase 1, DEP1, DEP-1, DEP1EC 3.1.3.48, HPTP eta, HPTPeta, HPTP-eta, human density enhanced phosphatase-1, protein tyrosine phosphatase, receptor type, J, protein tyrosine phosphatase, receptor type, J polypeptide, Protein-tyrosine phosphatase eta, Protein-tyrosine phosphatase receptor type J, PTPRJ, receptor-type tyrosine-protein phosphatase eta, R-PTP-eta, R-PTP-J, SCC1, susceptibility to colon cancer 1, mouse, homolog of | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human DEP-1/CD148. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction