missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human COX-1 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB3740V-100UG
This item is not returnable.
View return policy
Description
COX-1 Monoclonal specifically detects COX-1 in Human samples. It is validated for Flow Cytometry.
Specifications
| COX-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P23219 | |
| PTGS1 | |
| E. coli-derived recombinant human COX-1 Ala24-Leu599 Accession # P23219 | |
| RUO | |
| 5742 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 345305 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| COX1, COX-1, COX1PGG/HS, COX3, Cyclooxygenase-1, EC 1.14.99.1, PCOX1, PGH synthase 1, PGHS1, PGHS-1PHS1, PHS 1, prostaglandin G/H synthase 1, prostaglandin G/H synthase and cyclooxygenase, Prostaglandin H2 synthase 1, Prostaglandin-endoperoxide synthase 1, prostaglandin-endoperoxide synthase 1 (prostaglandin G/H synthase andcyclooxygenase), PTGHS, PTGS1 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects human COX-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Semaphorin 3A, 3B, 6B, 6C, 6D, recombinant mouse Semaphorin 3A, 3B, 3E, 3F, 6A, or 7A is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction