missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Claudin-10b Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB4279S-100UG
This item is not returnable.
View return policy
Description
Claudin-10b Monoclonal specifically detects Claudin-10b in Human samples. It is validated for Flow Cytometry.
Specifications
| Claudin-10b | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P78369 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Claudin-10b. Stains human Claudin-10b transfectants but not irrelevant transfectants. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 394513 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CLDN10 | |
| NS0 mouse myeloma cell line transfected with human Claudin-10 b Isoform Met1-Val228 Accession # P78369 | |
| RUO | |
| 9071 | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction