missing translation for 'onlineSavingsMsg'
Learn More

Human CD200 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Product Code. 18725609
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18725609 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18725609 Supplier R&D Systems Supplier No. FAB27241T100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD200/OX2 Monoclonal specifically detects CD200/OX2 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD200/OX2
Applications Flow Cytometry
Classification Monoclonal
Clone 325516
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P41217.3
Gene Alias antigen identified by monoclonal MRC OX-2, CD200, CD200 antigenMOX1, CD200 molecule, MOX1, MOX2, MOX2MRC, MRC, MRC OX-2 antigen, OX-2, OX-2 membrane glycoprotein
Gene Symbols CD205
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD200 Gln31-Gly232 Accession # P41217.3
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4345
Test Specificity Detects human CD200 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse CD200 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.