missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ALK-1 Antibody, R&D Systems™

Description
ALK-1 Monoclonal specifically detects ALK-1 in Human samples. It is validated for Western Blot, Simple Western.
Specifications
Specifications
| Antigen | ALK-1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 117720 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 ug/mL, Simple Western 40 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | P37023 |
| Gene Alias | activin A receptor type II-like 1, activin A receptor, type II-like kinase 1, Activin receptor-like kinase 1, ACVRL1, ACVRLK1ORW2, ALK1, ALK-1, ALK1TGF-B superfamily receptor type I, EC 2.7.11, HHT, HHT2EC 2.7.11.30, serine/threonine-protein kinase receptor R3, SKR3, TSR-I |
| Gene Symbols | ACVRL1 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?