missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ADAM10 Ectodomain Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC1427N-100UG
This item is not returnable.
View return policy
Description
ADAM10 Monoclonal specifically detects ADAM10 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| ADAM10 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| O14672 | |
| ADAM10 | |
| Mouse myeloma cell line NS0-derived recombinant human ADAM10 Thr214-Glu672 Accession # O14672 | |
| RUO | |
| 102 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 163003 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| a disintegrin and metalloprotease domain 10, a disintegrin and metalloproteinase domain 10, AD10, ADAM 10, ADAM metallopeptidase domain 10, CD156c, CD156c antigen, CDw156, disintegrin and metalloproteinase domain-containing protein 10, EC 3.4.24, HsT18717, kuz, Kuzbanian, Kuzbanian protein homolog, MADM, MADMEC 3.4.24.81, Mammalian disintegrin-metalloprotease | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human ADAM10 in direct ELISAs and Western blots. In Western blots, shows less than 5% cross-reactivity with recombinant human (rh) ADAM8, 9, 15, 17/TACE, and recombinant mouse ADAM10. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction