missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HA Tag Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6875
This item is not returnable.
View return policy
Description
HA Tag Monoclonal specifically detects HA Tag in Multi-Species samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| HA Tag | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HA, HA epitope tag, Hemagglutinin | |
| HA Tag peptide Accession # ABB51961 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 912426 | |
| Western Blot 0.5 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| ABB51961 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Detects N-terminal,C-terminal, and Internal Sequence HA-tagged proteins in Western blot. | |
| Multi-species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction