Learn More
Human/Primate CXCL1/GRO alpha/KC/CINC-1 Alexa Fluor 350-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems IC2751U-100UG
This item is not returnable.
View return policy
Description
CXCL1/GRO alpha/KC/CINC-1 Monoclonal specifically detects CXCL1/GRO alpha/KC/CINC-1 in Human, Primate samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| CXCL1/GRO alpha/KC/CINC-1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human and primate CXCL1/GRO alpha/KC/CINC-1 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 20326R | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1 | |
| E. coli-derived recombinant human CXCL1/GRO alpha/KC/CINC-1 | |
| RUO | |
| 2919 | |
| Human, Primate | |
| IgG2b |
Product Content Correction
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction