missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse RBP4/Retinol-Binding Protein 4 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB34761R-100UG
This item is not returnable.
View return policy
Description
RBP4/Retinol-Binding Protein 4 Monoclonal specifically detects RBP4/Retinol-Binding Protein 4 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| RBP4/Retinol-Binding Protein 4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_035385 | |
| RBP4 | |
| Mouse myeloma cell line NS0-derived recombinant mouse RBP4/Retinol-Binding Protein 4 Met1-Leu201 Accession # NP_035385 | |
| RUO | |
| 5950 | |
| Human, Mouse | |
| IgG2a |
| Flow Cytometry | |
| 423619 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, RetinolBinding Protein 4, retinol-binding protein 4, plasma | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse RBP4/Retinol-Binding Protein 4 in direct ELISA and Western Blot. In Western blots, detects human RBP4/Retinol-Binding Protein 4. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction