missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat HNF-3 beta/FoxA2 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems FAB24001R-100UG
This item is not returnable.
View return policy
Description
HNF-3 beta/FoxA2 Monoclonal specifically detects HNF-3 beta/FoxA2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| HNF-3 beta/FoxA2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| forkhead box A2, Forkhead box protein A2, FoxA2, hepatic nuclear factor-3-beta, hepatocyte nuclear factor 3, beta, hepatocyte nuclear factor 3-beta, HNF3 beta, HNF-3B, HNF-3-beta, HNF3BTCF-3B, MGC19807, TCF3B, Transcription factor 3B | |
| Human/Mouse/Rat HNF-3 beta/FoxA2 synthetic peptide Accession # Q9Y261 | |
| 100 μg | |
| 3170 | |
| Human, Mouse, Rat | |
| Supernatant |
| Flow Cytometry | |
| 2141A | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9Y261 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human HNF-3 beta/FoxA2 in Western blots. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction