Learn More
Tocris Bioscience™ GsMTx4
GREENER_CHOICE
Product Code. 18745727
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Product Code. 18745727
Brand: Tocris Bioscience™ 4912/100U
TRPC1 and TRPC6 inhibitor; inhibits mechanosensitive ion channels
GsMTx4 is a spider venom peptide that selectively inhibits cationic mechanosensitive channels (MSCs) such as TRPC1 and TRPC6 and Piezo channels. GsMTx4 blocks stretch-activated cation channels in astrocytes, cardiac cells, and smooth and skeletal muscle cells. GsMTx4 also inhibits TACAN, a mechanosensitive ion channel involved in the pain response. GsMTx4 decreases the leptin-induced AMPK and MLC-2 phosphorylation in breast epithelial cells. GsMTx4 is neuroprotective and inhibits lysophosphatidylcholine-induced astrocyte toxicity in vivo in mice. GsMTx4 suppresses neurogenesis and enhances astrogenesis in human neural stem cells.
TRUSTED_SUSTAINABILITY
Specifications
CAS 1209500-46-8