missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD3 PerCP-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems FAB4841C-100
This item is not returnable.
View return policy
Description
CD3 Monoclonal specifically detects CD3 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD3 | |
| Monoclonal | |
| PerCP | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Cd247 | |
| T cell hybridoma D1 | |
| 100 Tests | |
| Primary | |
| Reacts with mouse TCR-associated CD3 complex that occurs on thymocytes and mature T cells. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 17A2 | |
| Flow Cytometry 10 uL/10^6 cells | |
| CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, CD3G, CD3g antigen, CD3g antigen, gamma polypeptide (TiT3 complex), CD3g molecule, epsilon (CD3-TCR complex), CD3g molecule, gamma (CD3-TCR complex), CD3-GAMMA, FLJ17620, FLJ17664, FLJ18683, FLJ79544, FLJ94613, MGC138597, T3DOKT3, delta chain, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell receptor T3 delta chain, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 delta chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 916 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction