missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TGF-beta RII Fluorescein-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems FAB2411F-100
This item is not returnable.
View return policy
Description
TGF-beta RII Polyclonal specifically detects TGF-beta RII in Human samples. It is validated for Flow Cytometry.
Specifications
| TGF-beta RII | |
| Polyclonal | |
| Flow Cytometry 10 uL/10^6 cells | |
| P37173.2 | |
| TGFBR2 | |
| Mouse myeloma cell line NS0-derived recombinant human TGF-beta RII Ile24-Asp159 Accession # P37173.2 | |
| 100 Tests | |
| Primary | |
| Detects human TGF-beta RII in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse (rm) TGF-beta RII and less than 1% cross-reactivity with recombinant human TGF-beta RIII and rmTGF-beta RI is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| Flow Cytometry | |
| Fluorescein | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| AAT3, EC 2.7.11, EC 2.7.11.30, FAA3, HNPCC6, LDS1B, LDS2B, MFS2, RIIC, TAAD2, tbetaR-II, TGF-beta receptor type II, TGF-beta receptor type IIB, TGF-beta receptor type-2, TGF-beta type II receptor, TGFbetaRII, TGFbeta-RII, TGFBR2, TGF-bRII, TGFR-2, transforming growth factor beta receptor type IIC, transforming growth factor, beta receptor II (70/80kDa), transforming growth factor, beta receptor II (70/80kDa) isoform 1, transforming growth factor, beta receptor II (70/80kDa) isoform 2, transforming growth factor, beta receptor II (70-80kD), Transforming growth factor-beta receptor type II | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7048 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction