missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-18/IL-1F4 Propeptide PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems IC646P
This item is not returnable.
View return policy
Description
IL-18/IL-1F4 Monoclonal specifically detects IL-18/IL-1F4 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IL-18/IL-1F4 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Iboctadekin, IFN-gamma-inducing factor, IGIF, IGIFIL-1 gamma, IL18, IL-18MGC12320, IL-1F4, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3606 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 74801 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| Q14116 | |
| IL18 | |
| E. coli-derived recombinant human IL-18/IL-1F4 Accession # Q14116 | |
| 100 Tests | |
| Primary | |
| Detects the pro region of human IL-18/IL-1F4 in direct ELISAs and Western blots. Does not cross-react with recombinant human IL-18, recombinant mouse IL-18 or recombinant rat IL-18. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction