missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HLA-DR Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB4869R-100
This item is not returnable.
View return policy
Description
HLA-DR Monoclonal specifically detects HLA-DR in Human samples. It is validated for Flow Cytometry.
Specifications
| HLA-DR | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLADR, HLA-DRA, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MHC Class II DR, MLRW | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3122 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| L203 | |
| Flow Cytometry 5 uL/10^6 cells | |
| P01903 | |
| HLA-DRA | |
| RPMI 8866 human lymphoblastoid cells Accession # P01903 | |
| 100 Tests | |
| Primary | |
| Detects human HLA-DR. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction