missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tungsten Standard: W @ 10000 μg/mL in 5% HNO3, tr. HF, VHG™
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
Description
Tungsten Standard: W @ 10000 μg/mL in 5% HNO3, tr. HF
Specifications
Specifications
| Chemical Name or Material | Tungsten |
| CAS | 7440-33-7 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Tungsten |
| Shipping Condition | Room Temperature |
| Molecular Formula | W |
| Concentration | 10000 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | W |
| InChI Formula | InChI=1S/W |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction