missing translation for 'onlineSavingsMsg'
Learn More

Tellurium Standard: Te @ 10000 μg/mL in 30% HCl, VHG™

Product Code. 18053140
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
This item is not returnable. View return policy

Product Code. 18053140

Brand: VHG Labs VHGTTEH100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Tellurium Standard: Te @ 10000 μg/mL in 30% HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tellurium
CAS 13494-80-9
Methods ICP/ICP-MS
Analyte or Component Names Tellurium
Shipping Condition Room Temperature
Molecular Formula Te
Concentration 10000 μg/mL
Certifications/Compliance ISO 17034
Synonym Te
SMILES [Te]
IUPAC Name Tellurium
Solvent or Matrix Dilute HCl
Molecular Weight (g/mol) 127
Quantity 100 mL
Formula Weight 127.6 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.