missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sn @ 5μg/mL; Si @ 10μg/mL; Mo @ 20μg/mL in 3% HNO3 (v/v)/tr HF, VHG™
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
Description
Sn @ 5μg/mL; Si @ 10μg/mL; Mo @ 20μg/mL in 3% HNO3 (v/v)/tr HF
Specifications
Specifications
| Chemical Name or Material | Tin/Silicon/Molybdenum |
| CAS | 7440-31-5, 7440-21-3, 7439-98-7 |
| Analyte or Component Names | Tin/Silicon/Molybdenum |
| Shipping Condition | Room Temperature |
| Solvent or Matrix | 3% HNO3 |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Tin 5 μg/mL/Silicon 10 μg/mL/Molybdenum 20 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction