missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chem-Lab 100mL Silver standard solution 10 mg Ag / L 10% HN
Brand: Chem-Lab CL01.2601.010.0100
100mL Silver standard solution 10 mg Ag / L 10% HNO3
This item is not returnable.
View return policy
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Silver standard solution 10 µg/ml
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Nitric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction