missing translation for 'onlineSavingsMsg'
Learn More

Mo, Sn, Ti, Si @ 1000μg/mL; Matrix:5% HNO3.tr HF, 100mL , VHG™

Product Code. 17958419
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 17958419

Brand: VHG Labs VHG1101100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Mo, Sn, Ti, Si @ 1000μg/mL; Matrix:5% HNO3.tr HF, 100mL
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Molybdenum/Silicon/Tin/Titanium
CAS 7439-98-7, 7440-21-3, 7440-31-5, 7440-32-6
Analyte or Component Names Molybdenum/Silicon/Tin/Titanium
Shipping Condition Room Temperature
Solvent or Matrix 5% HNO3
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Molybdenum 1000 μg/mL/Silicon 1000 μg/mL/Tin 1000 μg/mL/Titanium 1000 μg/mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.