missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MISA Standard 6: Ag, Al, B, Ba, Ca, Cd, Co, Cr, Cs, Cu, Ga, In, Fe, K, Li, Mg, Mn, Na, Ni, P, Pb, Rb, Sr, Tl, U, V, Zn @ 100 μg/mL in 2% HNO3, VHG™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
MISA Standard 6: Ag, Al, B, Ba, Ca, Cd, Co, Cr, Cs, Cu, Ga, In, Fe, K, Li, Mg, Mn, Na, Ni, P, Pb, Rb, Sr, Tl, U, V, Zn @ 100 μg/mL in 2% HNO3
Specifications
Specifications
| Chemical Name or Material | Aluminium/Barium/Boron/Cadmium/Calcium/Cesium/Chromium/Cobalt/Copper/Gallium/Indium/Iron/Lead/Lithium/Magnesium/Manganese/Nickel/Phosphorus/Potassium/Rubidium/Silver/Sodium/Strontium/Thallium/Uranium/Vanadium/Zinc |
| CAS | 7429-90-5, 7440-39-3, 7440-42-8, 7440-43-9, 7440-70-2, 7440-46-2 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Aluminium/Barium/Boron/Cadmium/Calcium/Cesium/Chromium/Cobalt/Copper/Gallium/Indium/Iron/Lead/Lithium/Magnesium/Manganese/Nickel/Phosphorus/Potassium/Rubidium/Silver/Sodium/Strontium/Thallium/Uranium/Vanadium/Zinc |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Aluminium 100 μg/mL/Barium 100 μg/mL/Boron 100 μg/mL/Cadmium 100 μg/mL/Calcium 100 μg/mL/Cesium 100 μg/mL/Chromium 100 μg/mL/Cobalt 100 μg/mL/Copper 100 μg/mL/Gallium 100 μg/mL/Indium 100 μg/mL/Iron 100 μg/mL/Lead 100 μg/mL/Lithium 100 μg/mL/Magnesium 100 μg/mL/Manganese 100 μg/mL/Nickel 100 μg/mL/Phosphorus 100 μg/mL/Potassium 100 μg/mL/Rubidium 100 μg/mL/Silver 100 μg/mL/Sodium 100 μg/mL/Strontium 100 μg/mL/Thallium 100 μg/mL/Uranium 100 μg/mL/Vanadium 100 μg/mL/Zinc 100 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction