missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICP-MS Internal Standard: Sc @ 100 ug/mL in 2% HNO3, VHG™
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
ICP-MS Internal Standard: Sc @ 100 ug/mL in 2% HNO3
Specifications
Specifications
| Chemical Name or Material | Scandium |
| CAS | 7440-20-2 |
| Analyte or Component Names | Scandium |
| Shipping Condition | Room Temperature |
| Molecular Formula | Sc |
| Concentration | 100 μg/mL |
| Synonym | Sc |
| InChI Formula | InChI=1S/Sc |
| SMILES | [Sc] |
| IUPAC Name | scandium |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction