missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ag, Al, B, Ba, Bi, Ca, Cd, Co, Cr, Cu, Fe, Ga, In, K, Li, Mg, Mn, Na, Ni, Pb, Sr, Tl, Zn @ 1000 ug/mL in 5% HNO3, VHG™
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
Description
Ag, Al, B, Ba, Bi, Ca, Cd, Co, Cr, Cu, Fe, Ga, In, K, Li, Mg, Mn, Na, Ni, Pb, Sr, Tl, Zn @ 1000 ug/mL in 5% HNO3
Specifications
Specifications
| Chemical Name or Material | Aluminium/Barium/Bismuth/Boron/Cadmium/Calcium/Chromium/Cobalt/Copper/Gallium/Indium/Iron/Lead/Lithium/Magnesium/Manganese/Nickel/Potassium/Silver/Sodium/Strontium/Thallium/Zinc |
| CAS | 7429-90-5, 7440-39-3, 7440-69-9, 7440-42-8, 7440-43-9, 7440-70-2 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Aluminium/Barium/Bismuth/Boron/Cadmium/Calcium/Chromium/Cobalt/Copper/Gallium/Indium/Iron/Lead/Lithium/Magnesium/Manganese/Nickel/Potassium/Silver/Sodium/Strontium/Thallium/Zinc |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Aluminium 1000 μg/mL/Barium 1000 μg/mL/Bismuth 1000 μg/mL/Boron 1000 μg/mL/Cadmium 1000 μg/mL/Calcium 1000 μg/mL/Chromium 1000 μg/mL/Cobalt 1000 μg/mL/Copper 1000 μg/mL/Gallium 1000 μg/mL/Indium 1000 μg/mL/Iron 1000 μg/mL/Lead 1000 μg/mL/Lithium 1000 μg/mL/Magnesium 1000 μg/mL/Manganese 1000 μg/mL/Nickel 1000 μg/mL/Potassium 1000 μg/mL/Silver 1000 μg/mL/Sodium 1000 μg/mL/Strontium 1000 μg/mL/Thallium 1000 μg/mL/Zinc 1000 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction