missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gold Stabilizer for Mercury: Au @ 100 μg/mL in 5% HCl, VHG™
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
Gold Stabilizer for Mercury: Au @ 100 μg/mL in 5% HCl
Specifications
Specifications
| Chemical Name or Material | Gold |
| CAS | 7440-57-5 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Gold |
| Shipping Condition | Room Temperature |
| Molecular Formula | Au |
| Certifications/Compliance | ISO 17034 |
| Synonym | Colloidal gold, Gold powder, Gold flake, Gold leaf |
| InChI Formula | InChI=1S/Au |
| SMILES | [Au] |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction