Learn More
Fisher Chemical 100ml Formaldehyde Standard Solution 5 ppm CH2O

Brand: Fisher Chemical BPS 404
100ml Formaldehyde Standard Solution 5 ppm CH2O
Safety and Handling
- British Pharmacopoeia Formaldehyde Standard Solution
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1B
Hazard Statement
- H350-May cause cancer.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contain: formaldehyde
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.