Learn More
Reagecon 100ML ETALON DENSITE 0.8629G/ML @80°C
Brand: Reagecon DEN80120
100ML ETALON DENSITE 0.8629G/ML @80°C
Safety and Handling
- DENSITY STANDARD 0.8629g/ml @ 80°C
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Flammable liquid Category 3
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contain: pentanol isomers
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.