Learn More
Reagecon 100ML ETALON DENSITE 0.8084G/ML @25°C
Brand: Reagecon DEN25090
100ML ETALON DENSITE 0.8084G/ML @25°C
Safety and Handling
- DENSITY STANDARD 0.8084g/ml @ 25°C
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P321-Specific treatment (see on this label).
- P331-Do NOT induce vomiting.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contain: toluene, dodecane
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.