missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Standard: Al @ 10 μg/mL in 2% HNO3, VHG™

Product Code. 17963166
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17963166 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17963166 Supplier VHG Labs Supplier No. VHGLALN100

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Aluminum Standard: Al @ 10 μg/mL in 2% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminium
CAS 7429-90-5
Methods ICP/ICP-MS
Analyte or Component Names Aluminium
Shipping Condition Room Temperature
Molecular Formula Al
Concentration 10 μg/mL
Certifications/Compliance ISO 17034
Synonym Aluminium
InChI Formula InChI=1S/Al
SMILES [Al]
IUPAC Name Aluminum
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 26.98
Quantity 100 mL
Formula Weight 26.9815 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.