missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Al, Cu @ 2μg/mL; Mg, Zn @ 4μg/mL; Fe @ 5μg/mL; Ca, Pb @ 10μg/mL; B @ 50μg/mL; P @ 400μg/mL; K, Na @ 1000μg/mL in 3% HNO3 (v/v), VHG™
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
Description
Al, Cu @ 2μg/mL; Mg, Zn @ 4μg/mL; Fe @ 5μg/mL; Ca, Pb @ 10μg/mL; B @ 50μg/mL; P @ 400μg/mL; K, Na @ 1000μg/mL in 3% HNO3 (v/v)
Specifications
Specifications
| Chemical Name or Material | Aluminium/Copper/Magnesium/Zinc/Iron/Calcium/Lead/Boron/Phosphorus/Potassium/Sodium |
| CAS | 7429-90-5, 7440-50-8, 7439-95-4, 7440-66-6, 7439-89-6, 7440-70-2 |
| Analyte or Component Names | Aluminium/Copper/Magnesium/Zinc/Iron/Calcium/Lead/Boron/Phosphorus/Potassium/Sodium |
| Shipping Condition | Room Temperature |
| Solvent or Matrix | 3% HNO3 |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Aluminium 2 μg/mL/Copper 2 μg/mL/Magnesium 4 μg/mL/Zinc 4 μg/mL/Iron 5 μg/mL/Calcium 10 μg/mL/Lead 10 μg/mL/Boron 50 μg/mL/Phosphorus 400 μg/mL/Potassium 1000 μg/mL/Sodium 1000 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction