missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yohimbine N,O-Methoxycarbonate, TRC
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
Specifications
Specifications
| Chemical Name or Material | Yohimbine N,O-Methoxycarbonate |
| Quantity | 100 mg |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25 H30 N2 O7 |
| Formula Weight | 470.205 |
| Molecular Weight (g/mol) | 470.515 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction