missing translation for 'onlineSavingsMsg'
Learn More

XC-DAPOL-CPBA (>90%), TRC

Product Code. 30451647
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30451647

Brand: Toronto Research Chemicals TRCX742015100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Xylene cyanole-Dapol-phenylboronic Acid
CAS 191231-97-7
Molecular Formula C35H41BN4O9S2
Quantity 100 mg
Synonym XC-DAPOL-CPBA, 5-[[[3-[(4-Boronobenzoyl)amino]-2-hydroxypropyl]amino]sulfonyl]-2-[[4-(ethylamino)-3-methylphenyl][4-(ethylimino)-3-methyl-2,5-cyclohexadien-1-ylidene]methyl]benzenesulfonic Acid
InChI Formula InChI=1S/C35H41BN4O9S2/c1-5-37-31-15-9-25(17-22(31)3)34(26-10-16-32(38-6-2)23(4)18-26)30-14-13-29(19-33(30)51(47,48)49)50(45,46)40-21-28(41)20-39-35(42)24-7-11-27(12-8-24)36(43)44/h7-19,28,37,40-41,43-44H,5-6,20-21H2,1-4H3,(H,39,42)(H,47,48,49)
SMILES CCNc1ccc(cc1C)\C(=C\2/C=CC(=[NH+]CC)C(=C2)C)\c3ccc(cc3S(=O)(=O)[O-])S(=O)(=O)NCC(O)CNC(=O)c4ccc(cc4)B(O)O
IUPAC Name 5-(N-(3-(4-boronobenzamido)-2-hydroxypropyl)sulfamoyl)-2-((4-(ethylamino)-3-methylphenyl)(4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene)methyl)benzenesulfonic acid
Molecular Weight (g/mol) 736.66
Formula Weight 736.24
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.