missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Veratric Acid-d6, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Veratric Acid-d6 |
| CAS | 1162658-12-9 |
| Molecular Formula | C9H4D6O4 |
| Quantity | 100 mg |
| Synonym | 3,4-Dimethoxybenzoic Acid-d6, Dimethylprotocatechuic Acid-d6, NSC 7721-d6 |
| InChI Formula | InChI=1S/C9H10O4/c1-12-7-4-3-6(9(10)11)5-8(7)13-2/h3-5H,1-2H3,(H,10,11)/i1D3,2D3 |
| SMILES | [2H]C([2H])([2H])OC1=C(OC([2H])([2H])[2H])C=CC(C(O)=O)=C1 |
| Molecular Weight (g/mol) | 188.21 |
| Formula Weight | 188.0956 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction