missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vasopressin Acetic Acid Salt, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Vasopressin Acetic Acid Salt |
| Molecular Formula | C46H65N15O12S2 • (C2H4O2) |
| Quantity | 100 mg |
| Synonym | 1,2-Dithia-5,8,11,14,17-pentaazacycloeicosane','Cyclic Peptide Derivative Acetic Acid Salt, ADH Acetic Acid Salt, ADH (hormone) Acetic Acid Salt, Antidiuretic Hormone Acetic acid salt, Intertocin Acetic Acid Salt, Pituitrin P Acetic Acid Salt, Tonephin Acetic Acid Salt, Vasopressin Injection Acetic Acid Salt, Vasopressins Acetic Acid Salt, β-Hypophamine Acetic Acid Salt |
| SMILES | CC1=CC(C)=C(C(N1N)=O)C#N |
| Molecular Weight (g/mol) | 1084.236005 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction