missing translation for 'onlineSavingsMsg'
Learn More

Vardenafil N-Oxide, TRC

Product Code. 30449929
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30449929

Brand: Toronto Research Chemicals TRCV098010100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vardenafil N-Oxide
CAS 448184-48-3
Molecular Formula C23 H32 N6 O5 S
Quantity 100 mg
Synonym Imidazo[5,1-f][1,2,4]triazin-4(1H)-one','2-[2-ethoxy-5-[(4-ethyl-4-oxido-1-piperazinyl)sulfonyl]phenyl]-5-methyl-7-propyl-, Piperazine','1-[[3-(1,4-dihydro-5-methyl-4-oxo-7-propylimidazo[5,1-f][1,2,4]triazin-2-yl)-4-ethoxyphenyl]sulfonyl]-4-ethyl-','4-oxide (9CI), Vardenafil N-Oxide, 2-{2-Ethoxy-5-[(4-ethyl-4-oxidopiperazin-1-yl)sulfonyl]phenyl}-5-methyl-7-propyl-imidazo[5,1-f][1,2,4]triazin-4(3H)-one
InChI Formula InChI=1S/C23H32N6O5S/c1-5-8-20-24-16(4)21-23(30)25-22(26-28(20)21)18-15-17(9-10-19(18)34-7-3)35(32,33)27-11-13-29(31,6-2)14-12-27/h9-10,15H,5-8,11-14H2,1-4H3,(H,25,26,30)
SMILES CCCc1nc(C)c2C(=O)NC(=Nn12)c3cc(ccc3OCC)S(=O)(=O)N4CC[N+]([O-])(CC)CC4
IUPAC Name 2-[2-ethoxy-5-(4-ethyl-4-oxidopiperazin-4-ium-1-yl)sulfonylphenyl]-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one
Molecular Weight (g/mol) 504.6
Formula Weight 504.2155
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.