missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimeprazine Sulphoxide Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Trimeprazine Sulphoxide Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Trimeprazine Sulphoxide Hydrochloride |
| CAS | 2731414-34-7 |
| Analyte or Component Names | Trimeprazine Sulphoxide Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H22 N2 O S . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Trimeprazine Sulphoxide Hydrochloride |
| InChI Formula | InChI=1S/C18H22N2OS.ClH/c1-14(12-19(2)3)13-20-15-8-4-6-10-17(15)22(21)18-11-7-5-9-16(18)20;/h4-11,14H,12-13H2,1-3H3;1H |
| SMILES | Cl.CC(CN(C)C)CN1c2ccccc2S(=O)c3ccccc13 |
| IUPAC Name | N,N,2-trimethyl-3-(5-oxophenothiazin-10-yl)propan-1-amine;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction