missing translation for 'onlineSavingsMsg'
Learn More

TCS-PIM-1-4a, MedChemExpress

Product Code. 16437126
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16437126

Brand: Medchem Express HY16576

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress TCS-PIM-1-4a (SMI-4a) is a pan-Pim kinases inhibitor that blocks mTORC1 activity via activation of AMPK. TCS-PIM-1-4a kills a wide range of both myeloid and lymphoid cell lines (IC50 values ranging from 0.8 μM to 40 μM).

TCS-PIM-1-4a (SMI-4a) is a pan-Pim kinases inhibitor that blocks mTORC1 activity via activation of AMPK. TCS-PIM-1-4a kills a wide range of both myeloid and lymphoid cell lines (IC50 values ranging from 0.8 μM to 40 μM).

Specifications

Chemical Name or Material TCS-PIM-1-4a
Physical Form Solid
CAS 327033-36-3
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C11H6F3NO2S
For Use With (Application) Cancer-Kinase/protease
Synonym SMI-4a
Solubility Information DMSO : ≥ 100 mg/mL (365.99 mM)
SMILES O=C(NC/1=O)SC1=C\C2=CC=CC(C(F)(F)F)=C2
Molecular Weight (g/mol) 273.23
Formula Weight 273.23
Percent Purity 99.73%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.