missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 4-Aminobenzenesulfonate Dihydrate (Sulfanilic Acid Sodium Salt Dihydrate), Mikromol™
Description
Sodium 4-Aminobenzenesulfonate Dihydrate (Sulfanilic Acid Sodium Salt Dihydrate)
Specifications
Specifications
| Chemical Name or Material | Sodium 4-Aminobenzenesulfonate Dihydrate |
| CAS | 6106-22-5 |
| Analyte or Component Names | Sodium 4-Aminobenzenesulfonate Dihydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H6 N O3 S . Na . 2 H2 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4-Aminobenzenesulfonic Acid Sodium Salt Hydrate (1:1:2), Benzenesulfonic acid, 4-amino-, monosodium salt, dihydrate (9CI), Sulfanilic acid, monosodium salt, dihydrate (8CI), Sodium sulfanilate dihydrate, Sulfanilic Acid Sodium Salt Dihydrate, Mesalazine Impurity O as Sodium Salt Dihydrate, Sulfadiazine Impurity B as Dihydrate Sodium Salt, Sulfadimidine Impurity F as Dihydrate Sodium Salt, Sulfamethoxazole Impurity D as Dihydrate Sodium Salt |
| InChI Formula | InChI=1S/C6H7NO3S.Na.2H2O/c7-5-1-3-6(4-2-5)11(8,9)10;;;/h1-4H,7H2,(H,8,9,10);;2*1H2/q;+1;;/p-1 |
| SMILES | O.O.[Na+].Nc1ccc(cc1)S(=O)(=O)[O-] |
| IUPAC Name | sodium;4-aminobenzenesulfonate;hydrate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?